missing translation for 'onlineSavingsMsg'
Learn More

4-Methylumbelliferyl-β-D-galactopyranoside, Thermo Scientific Chemicals

Product Code. 10736252
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6160-78-7
Molecular Formula C16H18O8
Molecular Weight (g/mol) 338.31
MDL Number MFCD00036773,MFCD00063694
InChI Key YUDPTGPSBJVHCN-DZQJYWQESA-N
Synonym 4-methylumbelliferyl-beta-d-galactopyranoside, 4-methylumbelliferyl-galactopyranoside, 4-methylumbelliferyl beta-d-galactoside, 4-methylumbelliferyl b-d-galactoside, muga, 4-methylumbelliferyl beta-galactoside, 7-beta-d-galactopyranosyloxy-4-methyl-2h-1-benzopyran-2-one, 4-methylumbelliferyl b-d-galactopyranoside, 4-methyl-7-2s,3r,4s,5r,6r-3,4,5-trihydroxy-6-hydroxymethyl tetrahydropyran-2-yl oxy-chromen-2-one, 4-methyl-7-2s,3r,4s,5r,6r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxy chromen-2-one
PubChem CID 93577
IUPAC Name 4-methyl-7-[(2S,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxychromen-2-one
SMILES CC1=CC(=O)OC2=CC(O[C@@H]3O[C@H](CO)[C@H](O)[C@H](O)[C@H]3O)=CC=C12

Specifications

Melting Point 248°C
Color White
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 1 g
Beilstein 18,V,1,444
Specific Rotation Condition −64° (20°C c=1,pyridine,24h)
Specific Rotation -64
Solubility Information Solubility in water: moderately soluble. Other solubilities: soluble in dmf,pyridine,moderately soluble in ethanol,dmso
Formula Weight 338.31
Percent Purity Complies
Physical Form Powder
Chemical Name or Material 4-Methylumbelliferyl-β-D-galactopyranoside
Show More Show Less
Product Identifier
  • 4-Methylumbelliferyl-ß-D-galactopyranoside

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'