Learn More
4-Morpholineacetic acid hydrochloride, 95%, Thermo Scientific Chemicals
POA - £130.65
Chemical Identifiers
CAS | 89531-58-8 |
---|---|
Molecular Formula | C6H12ClNO3 |
Molecular Weight (g/mol) | 181.62 |
MDL Number | MFCD03540888 |
InChI Key | ICGYBJNONRDZOP-UHFFFAOYSA-N |
Synonym | 2-morpholinoacetic acid hydrochloride, 2-morpholin-4-yl acetic acid hydrochloride, morpholin-4-ylacetic acid hydrochloride, morpholin-4-yl-acetic acid hydrochloride, 4-morpholinacetic acid hcl, 4-morpholineacetic acid hydrochloride, 4-morpholineaceticacid, hydrochloride, 4-morpholinylacetic acid hydrochloride, 4-morpholineacetic acid, hydrochloride, 4-morpholineaceticacid, hydrochloride 1:1 |
PubChem CID | 17750946 |
IUPAC Name | 2-morpholin-4-ylacetic acid;hydrochloride |
SMILES | [H+].[Cl-].OC(=O)CN1CCOCC1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15417698
|
Thermo Scientific Alfa Aesar
H61188.MD |
250 mg | N/A | ||||||
15497688
|
Thermo Scientific Alfa Aesar
H61188.03 |
1 g |
£30.35
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15407698
|
Thermo Scientific Alfa Aesar
H61188.06 |
5 g |
£130.65
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
4-Morpholineacetic acid hydrochloride is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Morpholineacetic acid hydrochloride is used as a pharmaceutical intermediate.
Solubility
Insoluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Chemical Identifiers
89531-58-8 | |
181.62 | |
ICGYBJNONRDZOP-UHFFFAOYSA-N | |
17750946 | |
[H+].[Cl-].OC(=O)CN1CCOCC1 |
C6H12ClNO3 | |
MFCD03540888 | |
2-morpholinoacetic acid hydrochloride, 2-morpholin-4-yl acetic acid hydrochloride, morpholin-4-ylacetic acid hydrochloride, morpholin-4-yl-acetic acid hydrochloride, 4-morpholinacetic acid hcl, 4-morpholineacetic acid hydrochloride, 4-morpholineaceticacid, hydrochloride, 4-morpholinylacetic acid hydrochloride, 4-morpholineacetic acid, hydrochloride, 4-morpholineaceticacid, hydrochloride 1:1 | |
2-morpholin-4-ylacetic acid;hydrochloride |
Specifications
89531-58-8 | |
C6H12ClNO3 | |
3701710 | |
Insoluble in water. | |
[H+].[Cl-].OC(=O)CN1CCOCC1 | |
181.62 | |
181.62 | |
Powder |
White | |
MFCD03540888 | |
2-morpholinoacetic acid hydrochloride, 2-morpholin-4-yl acetic acid hydrochloride, morpholin-4-ylacetic acid hydrochloride, morpholin-4-yl-acetic acid hydrochloride, 4-morpholinacetic acid hcl, 4-morpholineacetic acid hydrochloride, 4-morpholineaceticacid, hydrochloride, 4-morpholinylacetic acid hydrochloride, 4-morpholineacetic acid, hydrochloride, 4-morpholineaceticacid, hydrochloride 1:1 | |
ICGYBJNONRDZOP-UHFFFAOYSA-N | |
2-morpholin-4-ylacetic acid;hydrochloride | |
17750946 | |
95% | |
4-Morpholineacetic acid hydrochloride |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.