missing translation for 'onlineSavingsMsg'
Learn More

4-(Morpholinomethyl)aniline, 95+%, Thermo Scientific™

Product Code. 10517863
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 51013-67-3
Molecular Formula C11H16N2O
Molecular Weight (g/mol) 192.262
InChI Key WNYFVEFUHMDIRQ-UHFFFAOYSA-N
Synonym 4-morpholinomethyl aniline, 4-morpholin-4-ylmethyl aniline, 4-morpholin-4-ylmethylphenylamine, 4-morpholin-4-ylmethyl-phenylamine, benzenamine, 4-4-morpholinylmethyl, 4-4-morpholinylmethyl aniline, 4-morpholin-4-ylmethyl-aniline, 4-morpholin-4-ylmethyl phenylamine, acmc-209krj, 4-morpholinomethyl-aniline
PubChem CID 776851
IUPAC Name 4-(morpholin-4-ylmethyl)aniline
SMILES C1COCCN1CC2=CC=C(C=C2)N

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 192.26
Percent Purity 95%
Chemical Name or Material 4-(Morpholinomethyl)aniline
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'