missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(Morpholinomethyl)aniline, 95+%, Thermo Scientific™
£8.15 - £70.20
Chemical Identifiers
CAS | 51013-67-3 |
---|---|
Molecular Formula | C11H16N2O |
Molecular Weight (g/mol) | 192.262 |
InChI Key | WNYFVEFUHMDIRQ-UHFFFAOYSA-N |
Synonym | 4-morpholinomethyl aniline, 4-morpholin-4-ylmethyl aniline, 4-morpholin-4-ylmethylphenylamine, 4-morpholin-4-ylmethyl-phenylamine, benzenamine, 4-4-morpholinylmethyl, 4-4-morpholinylmethyl aniline, 4-morpholin-4-ylmethyl-aniline, 4-morpholin-4-ylmethyl phenylamine, acmc-209krj, 4-morpholinomethyl-aniline |
PubChem CID | 776851 |
IUPAC Name | 4-(morpholin-4-ylmethyl)aniline |
SMILES | C1COCCN1CC2=CC=C(C=C2)N |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10550423
|
Thermo Scientific Maybridge
CC29314CB |
250 mg | Amber Glass Bottle |
£8.15
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10517863
|
Thermo Scientific Maybridge
CC29314DA |
1 g | Amber Glass Bottle |
£18.40
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10357304
|
Thermo Scientific Maybridge
CC29314DE |
5 g | Amber Glass Bottle |
£70.20
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
51013-67-3 | |
192.262 | |
4-morpholinomethyl aniline, 4-morpholin-4-ylmethyl aniline, 4-morpholin-4-ylmethylphenylamine, 4-morpholin-4-ylmethyl-phenylamine, benzenamine, 4-4-morpholinylmethyl, 4-4-morpholinylmethyl aniline, 4-morpholin-4-ylmethyl-aniline, 4-morpholin-4-ylmethyl phenylamine, acmc-209krj, 4-morpholinomethyl-aniline | |
4-(morpholin-4-ylmethyl)aniline |
C11H16N2O | |
WNYFVEFUHMDIRQ-UHFFFAOYSA-N | |
776851 | |
C1COCCN1CC2=CC=C(C=C2)N |
Specifications
51013-67-3 | |
C11H16N2O | |
WNYFVEFUHMDIRQ-UHFFFAOYSA-N | |
4-(morpholin-4-ylmethyl)aniline | |
776851 | |
95% |
95% | |
4-morpholinomethyl aniline, 4-morpholin-4-ylmethyl aniline, 4-morpholin-4-ylmethylphenylamine, 4-morpholin-4-ylmethyl-phenylamine, benzenamine, 4-4-morpholinylmethyl, 4-4-morpholinylmethyl aniline, 4-morpholin-4-ylmethyl-aniline, 4-morpholin-4-ylmethyl phenylamine, acmc-209krj, 4-morpholinomethyl-aniline | |
C1COCCN1CC2=CC=C(C=C2)N | |
192.262 | |
192.26 | |
4-(Morpholinomethyl)aniline |