missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(Morpholinomethyl)benzoic acid, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
10 g
Packaging:
Amber Glass Bottle
Specifications
Specifications
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 250 mg |
Formula Weight | 221.25 |
Percent Purity | 97% |
Chemical Name or Material | 4-(Morpholinomethyl)benzoic acid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'