missing translation for 'onlineSavingsMsg'
Learn More

4-Morpholinophenylboronic acid, 97%, Thermo Scientific™

Product Code. 10279722
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 186498-02-2
Molecular Formula C10H14BNO3
Molecular Weight (g/mol) 207.04
MDL Number MFCD03095169
InChI Key WHDIUBHAKZDSJL-UHFFFAOYSA-N
Synonym 4-morpholinophenylboronic acid, 4-morpholinophenyl boronic acid, 4-morpholino phenylboronic acid, 4-morpholin-4-yl-phenylboronic acid, 4-morpholin-4-ylphenyl boronic acid, 4-4-boronophenyl morpholine, 4-morpholino phenyl boronic acid, 4-morpholin-4-yl phenylboronic acid, boronic acid, 4-4-morpholinyl phenyl, boronic acid, b-4-4-morpholinyl phenyl
PubChem CID 2795359
IUPAC Name (4-morpholin-4-ylphenyl)boronic acid
SMILES OB(O)C1=CC=C(C=C1)N1CCOCC1

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 207.04
Percent Purity 97%
Chemical Name or Material 4-Morpholinophenylboronic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.