missing translation for 'onlineSavingsMsg'
Learn More

4-n-Butylphenyl isocyanate, 98%

Product Code. 11362367
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11362367

Brand: Thermo Scientific Alfa Aesar L09957.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 69342-47-8 | C11H13NO | 175.231 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69342-47-8
Molecular Formula C11H13NO
Molecular Weight (g/mol) 175.231
MDL Number MFCD00013882
InChI Key LJJRXPXDTAUVQU-UHFFFAOYSA-N
Synonym 4-butylphenyl isocyanate, 4-n-butylphenyl isocyanate, benzene, 1-butyl-4-isocyanato, benzene,1-butyl-4-isocyanato, 4-butylbenzenisocyanate, 4-butylphenylisocyanate, acmc-209o7f, 1-butyl-4-isocyanato-benzene, 4-n-butyl phenyl isocyanate
PubChem CID 2796313
IUPAC Name 1-butyl-4-isocyanatobenzene
SMILES CCCCC1=CC=C(C=C1)N=C=O

Specifications

Density 0.992
Boiling Point 107°C to 108°C (3 mmHg)
Flash Point 108°C (226°F)
Refractive Index 1.5165
Quantity 1 g
UN Number UN2206
Beilstein 390859
Sensitivity Moisture sensitive
Formula Weight 175.23
Percent Purity 98%
Chemical Name or Material 4-n-Butylphenyl isocyanate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.