missing translation for 'onlineSavingsMsg'
Learn More

4-n-Decylbenzoyl chloride, 98%, Thermo Scientific Chemicals

Product Code. 11443457
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Employed as a pharmaceutical and chemical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Employed as a pharmaceutical and chemical intermediate.

Solubility
Reacts with water.

Notes
Moisture Sensitive. Store away from oxidizing agents, water/moisture. Store under inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54256-43-8
Molecular Formula C17H25ClO
Molecular Weight (g/mol) 280.836
MDL Number MFCD00041724
InChI Key HEWLDHSFOQXCSI-UHFFFAOYSA-N
PubChem CID 104714
IUPAC Name 4-decylbenzoyl chloride
SMILES CCCCCCCCCCC1=CC=C(C=C1)C(=O)Cl

Specifications

Boiling Point 171°C to 173°C (2 mmHg)
Flash Point >110°C (230°F)
Quantity 5 g
UN Number UN3265
Sensitivity Moisture sensitive
Synonym 4-n-decylbenzoyl chloride, benzoyl chloride, 4-decyl, p-decylbenzoyl chloride, 4-decylbenzoylchloride, p-n-decylbenzoyl chloride, benzoyl chloride,4-decyl, 4-n-decyl benzoyl chloride, 4-decylbenzoic acid chloride
Solubility Information Reacts with water.
Formula Weight 280.84
Percent Purity 98%
Chemical Name or Material 4-n-Decylbenzoyl chloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'