missing translation for 'onlineSavingsMsg'
Learn More

4-n-Decylbiphenyl, 98%, Thermo Scientific™

Product Code. 11452678
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
PDLC monomer. As chemical reagent.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 93972-02-2
Molecular Formula C22H30
Molecular Weight (g/mol) 294.482
MDL Number MFCD00102117
InChI Key YYBDFGOLQQVHDV-UHFFFAOYSA-N
Synonym 4-n-decylbiphenyl, 4-decyl-biphenyl, 4-decyl-1,1'-biphenyl, 4-decylbiphenyl, 4-decyl-1-phenylbenzene, maybridge1_002181, 1,1'-biphenyl, 4-decyl
PubChem CID 2801612
IUPAC Name 1-decyl-4-phenylbenzene
SMILES CCCCCCCCCCC1=CC=C(C=C1)C2=CC=CC=C2

Specifications

Melting Point 52°C to 54°C
Boiling Point 204°C to 210°C (1.5 mmHg)
Quantity 1 g
Solubility Information Insoluble in water.
Formula Weight 294.48
Percent Purity 98%
Chemical Name or Material 4-n-Decylbiphenyl
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'