missing translation for 'onlineSavingsMsg'
Learn More

4-[N-Methyl-N-(2-nitrophenyl)sulfamoyl]benzeneboronic acid, 97%, Thermo Scientific™

Product Code. 15401436
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1449133-61-2
Molecular Formula C13H13BN2O6S
Molecular Weight (g/mol) 336.13
MDL Number MFCD20265307
InChI Key LXCUTVNBMYMJPZ-UHFFFAOYSA-N
Synonym 4-n-methyl-n-2-nitrophenyl sulfamoyl benzeneboronic acid, 4-methyl 2-nitrophenyl sulfamoyl phenylboronic acid
PubChem CID 73995905
IUPAC Name [4-[methyl-(2-nitrophenyl)sulfamoyl]phenyl]boronic acid
SMILES CN(C1=CC=CC=C1[N+]([O-])=O)S(=O)(=O)C1=CC=C(C=C1)B(O)O

Specifications

Quantity 1 g
Formula Weight 336.13
Percent Purity 97%
Chemical Name or Material 4-[N-Methyl-N-(2-nitrophenyl)sulfamoyl]benzeneboronic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'