missing translation for 'onlineSavingsMsg'
Learn More

4-n-Octyloxybenzaldehyde, 97%, Thermo Scientific Chemicals

Product Code. 11425536
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24083-13-4
Molecular Formula C15H22O2
Molecular Weight (g/mol) 234.339
MDL Number MFCD00014136
InChI Key KVOWZHASDIKNFK-UHFFFAOYSA-N
Synonym 4-octyloxy benzaldehyde, 4-octyloxybenzaldehyde, 4-n-octyloxybenzaldehyde, p-octyloxybenzaldehyde, benzaldehyde, 4-octyloxy, p-octyloxy benzaldehyde, p-n-octoxy benzaldehyde, p-n-octyloxybenzaldehyde, acmc-209g8t
PubChem CID 90358
IUPAC Name 4-octoxybenzaldehyde
SMILES CCCCCCCCOC1=CC=C(C=C1)C=O

Specifications

Density 0.982
Boiling Point 140°C to 142°C (0.1 mmHg)
Refractive Index 1.522
Quantity 100 g
Beilstein 1913284
Sensitivity Air Sensitive
Formula Weight 234.33
Percent Purity 97%
Chemical Name or Material 4-n-Octyloxybenzaldehyde

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'