missing translation for 'onlineSavingsMsg'
Learn More

4-n-Octylphenol, 99%

Product Code. 11461340
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11461340

Brand: Thermo Scientific Alfa Aesar H26396.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1806-26-4 | C14H22O | 206.33 g/mol

4-n-Octylphenol is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-n-Octylphenol is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1806-26-4
Molecular Formula C14H22O
Molecular Weight (g/mol) 206.33
MDL Number MFCD00036134
InChI Key NTDQQZYCCIDJRK-UHFFFAOYSA-N
Synonym 4-n-octylphenol, phenol, 4-octyl, p-octylphenol, phenol, p-octyl, 1-p-hydroxyphenyl octane, para-octylphenol, unii-7df2b8lh3p, 4-octyl-phenol, 7df2b8lh3p, phenol, octenylated
PubChem CID 15730
ChEBI CHEBI:34432
IUPAC Name 4-octylphenol
SMILES CCCCCCCCC1=CC=C(O)C=C1

Specifications

Melting Point 43°C to 45°C
Density 0.961
Boiling Point 150°C (4 mmHg)
Flash Point >110°C (230°F)
Quantity 1 g
UN Number UN2430
Solubility Information Insoluble in water.
Formula Weight 206.32
Percent Purity 99%
Chemical Name or Material 4-n-Octylphenol
compliance-icons
Product Identifier
  • 4-n-Octylphenol
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : 4-n-Octylphenol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.