missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-n-Propoxyaniline, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
4-n-Propoxyaniline is used as a reactant for preparation of aggrecanase inhibitors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-n-Propoxyaniline is used as a reactant for preparation of aggrecanase inhibitors.
Solubility
Slightly soluble (1.3 g/L at 25°C).
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
4-n-Propoxyaniline is used as a reactant for preparation of aggrecanase inhibitors.
Solubility
Slightly soluble (1.3 g/L at 25°C).
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications
Specifications
Color | Brown |
Flash Point | 110°C (230°F) |
Quantity | 5 g |
Solubility Information | Slightly soluble (1.3g/L at 25°C). |
Formula Weight | 151.21 |
Percent Purity | 97% |
Physical Form | Liquid |
Chemical Name or Material | 4-n-Propoxyaniline |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'