missing translation for 'onlineSavingsMsg'
Learn More

4-n-Propoxybenzoic acid, 98%, Thermo Scientific Chemicals

Product Code. 11461497
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5438-19-7
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.20
MDL Number MFCD00013989
InChI Key GDFUWFOCYZZGQU-UHFFFAOYSA-N
Synonym p-propoxybenzoic acid, 4-n-propoxybenzoic acid, benzoic acid, 4-propoxy, 4-n-propyloxybenzoic acid, 4-propoxy-benzoic acid, benzoic acid, p-propoxy, p-propyloxybenzoic acid, pubchem2681, 4-propoxy benzoic acid, p-carboxyphenoxy propane
PubChem CID 138500
IUPAC Name 4-propoxybenzoic acid
SMILES CCCOC1=CC=C(C=C1)C(O)=O

Specifications

Melting Point 145°C to 147°C
Quantity 100 g
Beilstein 2208387
Formula Weight 180.2
Percent Purity 98%
Chemical Name or Material 4-n-Propoxybenzoic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'