missing translation for 'onlineSavingsMsg'
Learn More
4-Nitro-1H-pyrazole-3-carboxylic acid, 98%, Thermo Scientific Chemicals Product Code.: 11366659

4-Nitro-1H-pyrazole-3-carboxylic acid, 98%, Thermo Scientific Chemicals

Product Code. 11366659
10 g
Click to view available options
Quantity:
1 g
10 g
This item is not returnable. View return policy

Product Code. 11366659

Brand: Thermo Scientific Alfa Aesar H26224.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5334-40-7
Molecular Formula C4H3N3O4
Molecular Weight (g/mol) 157.085
MDL Number MFCD00090899
InChI Key ZMAXXOYJWZZQBK-UHFFFAOYSA-N
Synonym 4-nitro-1h-pyrazole-3-carboxylic acid, 4-nitropyrazole-3-carboxylic acid, 4-nitro-3-pyrazolecarboxylic acid, 4-nitro-1h-pyrazole-5-carboxylic acid, 1h-pyrazole-3-carboxylic acid, 4-nitro, 4-nitro-2h-pyrazole-3-carboxylic acid, 4-nitro pyrazole-3-carboxylic acid, 4-nitro-pyrazole-3-carboxylic acid, 4-hydroxynitroso-1h-pyrazole-3-carboxylic acid, nitropyrazolecarboxylic
PubChem CID 219739
SMILES C1=NNC(=C1[N+](=O)[O-])C(=O)O

Specifications

Melting Point ∼195°C (decomposition)
Quantity 10 g
Formula Weight 157.08
Percent Purity 98%
Chemical Name or Material 4-Nitro-1H-pyrazole-3-carboxylic acid

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10138492 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH26224.09
Lot Number10212944 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH26224.09
Lot Number10139718 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH26224.09
Lot Number10182698 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH26224.09
Lot Number10192282 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH26224.09

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Nitro-1H-pyrazole-3-carboxylic acid, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.