missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-nitro-L-phenylalanine, Thermo Scientific™
£16.15 - £140.00
Chemical Identifiers
CAS | 949-99-5 |
---|---|
Molecular Formula | C9H10N2O4 |
Molecular Weight (g/mol) | 210.19 |
InChI Key | GTVVZTAFGPQSPC-QMMMGPOBSA-N |
Synonym | 4-nitro-l-phenylalanine, h-phe 4-no2-oh, s-2-amino-3-4-nitrophenyl propanoic acid, 4-nitro-3-phenyl-l-alanine, l-4-nitrophenylalanine, p-nitro-l-phenylalanine, 2s-2-amino-3-4-nitrophenyl propanoic acid, p-nitrophenylalanine, l-phenylalanine, 4-nitro, l-4-no2-phe-oh |
PubChem CID | 13706 |
IUPAC Name | (2S)-2-amino-3-(4-nitrophenyl)propanoic acid |
SMILES | N[C@@H](CC1=CC=C(C=C1)[N+]([O-])=O)C(O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10630095
|
Thermo Scientific Maybridge
RJC02057DA |
1 g |
£16.15
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10376764
|
Thermo Scientific Maybridge
RJC02057FA |
100 g |
£140.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
949-99-5 | |
210.19 | |
4-nitro-l-phenylalanine, h-phe 4-no2-oh, s-2-amino-3-4-nitrophenyl propanoic acid, 4-nitro-3-phenyl-l-alanine, l-4-nitrophenylalanine, p-nitro-l-phenylalanine, 2s-2-amino-3-4-nitrophenyl propanoic acid, p-nitrophenylalanine, l-phenylalanine, 4-nitro, l-4-no2-phe-oh | |
(2S)-2-amino-3-(4-nitrophenyl)propanoic acid |
C9H10N2O4 | |
GTVVZTAFGPQSPC-QMMMGPOBSA-N | |
13706 | |
N[C@@H](CC1=CC=C(C=C1)[N+]([O-])=O)C(O)=O |
Specifications
949-99-5 | |
C9H10N2O4 | |
GTVVZTAFGPQSPC-QMMMGPOBSA-N | |
(2S)-2-amino-3-(4-nitrophenyl)propanoic acid | |
13706 | |
97% |
97% | |
4-nitro-l-phenylalanine, h-phe 4-no2-oh, s-2-amino-3-4-nitrophenyl propanoic acid, 4-nitro-3-phenyl-l-alanine, l-4-nitrophenylalanine, p-nitro-l-phenylalanine, 2s-2-amino-3-4-nitrophenyl propanoic acid, p-nitrophenylalanine, l-phenylalanine, 4-nitro, l-4-no2-phe-oh | |
N[C@@H](CC1=CC=C(C=C1)[N+]([O-])=O)C(O)=O | |
210.19 | |
210.19 | |
2-Amino-3-(4-nitrophenyl)propanoic acid |