missing translation for 'onlineSavingsMsg'
Learn More

4'-Nitroacetophenone, 98%, Thermo Scientific Chemicals

Product Code. 11411568
Click to view available options
Quantity:
50 g
250 g
1000 g
This item is not returnable. View return policy
This item is not returnable. View return policy

4'-Nitroacetophenone is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4′-Nitroacetophenone is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals.

Solubility
Insoluble in water.

Notes
Store in a cool, dry, well-ventilated area. Store away from oxidizing agent, strong alkalis.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-19-6
Molecular Formula C8H7NO3
Molecular Weight (g/mol) 165.15
MDL Number MFCD00007355
InChI Key YQYGPGKTNQNXMH-UHFFFAOYSA-N
Synonym p-nitroacetophenone, 4'-nitroacetophenone, 4-nitroacetophenone, 1-4-nitrophenyl ethanone, p-acetylnitrobenzene, paranitroacetophenone, p-nitrophenyl methyl ketone, 1-4-nitrophenyl ethan-1-one, ethanone, 1-4-nitrophenyl, pnap
PubChem CID 7487
ChEBI CHEBI:28735
IUPAC Name 1-(4-nitrophenyl)ethanone
SMILES CC(=O)C1=CC=C(C=C1)[N+]([O-])=O

Specifications

Melting Point 77°C to 81°C
Boiling Point 201°C to 202°C
Flash Point 149°C (300°F)
Assay Percent Range 98%
Odor Characteristic
Quantity 1000 g
Beilstein 607777
Solubility Information Insoluble in water.
Formula Weight 165.15
Percent Purity 98%
Chemical Name or Material 4'-Nitroacetophenone
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'