missing translation for 'onlineSavingsMsg'
Learn More
4-Nitroaniline, 98%, Thermo Scientific Chemicals Product Code.: 11414183

4-Nitroaniline, 98%, Thermo Scientific Chemicals

Product Code. 11414183
250 g
Click to view available options
Quantity:
250 g
1000 g
5000 g
This item is not returnable. View return policy

Product Code. 11414183

Brand: Thermo Scientific Alfa Aesar A10369.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

p-Nitroaniline is a a common chromophore and potentially useful syntetic reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
p-Nitroaniline is a a common chromophore and potentially useful syntetic reagent.

Solubility
Soluble in hot water and alcohol

Notes
Store at 4°C. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-01-6
Molecular Formula C6H6N2O2
Molecular Weight (g/mol) 138.13
MDL Number MFCD00007858
InChI Key TYMLOMAKGOJONV-UHFFFAOYSA-N
Synonym p-nitroaniline, p-nitraniline, 4-nitrobenzenamine, p-aminonitrobenzene, 4-nitraniline, benzenamine, 4-nitro, p-nitrophenylamine, 1-amino-4-nitrobenzene, aniline, p-nitro, developer pShow MoreShow Less
PubChem CID 7475
ChEBI CHEBI:17064
IUPAC Name 4-nitroaniline
SMILES NC1=CC=C(C=C1)[N+]([O-])=O

Specifications

Melting Point 147°C to 149°C
Density 1.437
Boiling Point 260°C (100 mmHg)
Flash Point 165°C (329°F)
Odor Amine-like
Quantity 250 g
UN Number UN1661
Beilstein 508690
Merck Index 14,6584
Solubility Information Soluble in hot water and alcohol
Formula Weight 138.13
Percent Purity 98%
Chemical Name or Material 4-Nitroaniline
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10249679 Certificate TypeCertificate of Analysis Date16/02/2024 Catalog NumberA10369.0B,A1036902,A1036901,A10369.30,A10369.0I
Lot Number10241655 Certificate TypeCertificate of Analysis Date04/07/2022 Catalog NumberA10369.0B,A10369.30,A10369.0I
Lot Number10118437 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA10369.0B,A10369.30,A10369.0I
Lot Number10210194 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA10369.0B,A10369.30,A10369.0I
Lot Number10175852 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA10369.0B,A10369.30,A10369.0I
compliance-icons compliance-icons
Product Identifier
  • 4-Nitroaniline
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H373-May cause damage to organs.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : 4-Nitroaniline

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Nitroaniline, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.