missing translation for 'onlineSavingsMsg'
Learn More

4-Nitroaniline, ≥99.0% (HPLC), Honeywell™ Fluka™

Product Code. 15696900
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Matrix substance for MALDI-MS,≥99.0% (HPLC)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-01-6
Molecular Formula C6H6N2O2
Molecular Weight (g/mol) 138.13
MDL Number MFCD00007858
InChI Key TYMLOMAKGOJONV-UHFFFAOYSA-N
PubChem CID 7475
ChEBI CHEBI:17064
IUPAC Name 4-nitroaniline
SMILES NC1=CC=C(C=C1)[N+]([O-])=O

Specifications

Melting Point 146°C to 149°C
Color Yellow
Density 1.44 g/cm3
pH 7
Boiling Point 260°C
Packaging Glass bottle
Linear Formula O2NC6H4NH2
Quantity 25 g
UN Number UN1661
Beilstein 508690
Formula Weight 138.12g/mol
Percent Purity ≥99.0% (HPLC)
Physical Form Crystals
Chemical Name or Material 4-Nitroaniline
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'