missing translation for 'onlineSavingsMsg'
Learn More
4-Nitroaniline, 99%, Thermo Scientific Chemicals Product Code.: 10657794

4-Nitroaniline, 99%, Thermo Scientific Chemicals

Product Code. 10657794
100 g, Glass bottle
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic Bottle
This item is not returnable. View return policy

Product Code. 10657794

Brand: Thermo Scientific Acros 128371000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-01-6
Molecular Formula C6H6N2O2
Molecular Weight (g/mol) 138.13
MDL Number MFCD00007858
InChI Key TYMLOMAKGOJONV-UHFFFAOYSA-N
Synonym p-nitroaniline, p-nitraniline, 4-nitrobenzenamine, p-aminonitrobenzene, 4-nitraniline, benzenamine, 4-nitro, p-nitrophenylamine, 1-amino-4-nitrobenzene, aniline, p-nitro, developer pShow MoreShow Less
PubChem CID 7475
ChEBI CHEBI:17064
IUPAC Name 4-nitroaniline
SMILES NC1=CC=C(C=C1)[N+]([O-])=O

Specifications

Melting Point 146.0°C to 151.0°C
Color Yellow to Brown
Boiling Point 332.0°C
Flash Point 199°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula O2NC6H4NH2
Quantity 100 g
Beilstein 12,711
Merck Index 15,667
Solubility Information Solubility in water: 0.8g/L (20°C). Other solubilities: soluble in alcohol,ether,benzene and methanol,1 g soluble in 25 mL alcohol,1 g soluble in 30 mL ether
Formula Weight 138.13
Percent Purity 99%
Physical Form Crystalline Powder, Crystals or Flakes
Chemical Name or Material 4-Nitroaniline
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0467376 Certificate TypeCertificate of Analysis Date30/01/2025 Catalog Number128375000,128370025,128371000
Lot NumberA0455271 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number128375000,128370025,128371000
Lot NumberA0449158 Certificate TypeCertificate of Analysis Date06/01/2023 Catalog Number128375000,128370025,128371000
Lot NumberA0400870 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number128375000,128370025,128371000
Lot NumberA0414497 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number128375000,128370025,128371000
compliance-icons compliance-icons
Product Identifier
  • 4-Nitroaniline
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Specific target organ toxicity after repeated exposure Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H373-May cause damage to organs.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Nitroaniline, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.