missing translation for 'onlineSavingsMsg'
Learn More

4-Nitroaniline, Pure, Fisher Chemical

Catalog Number 10538250
Click to view available options
Quantity:
100 g
Packaging:
Glass Jar
This item is not returnable. View return policy
This item is not returnable. View return policy
CAS 100-01-6
Melting Point 146°C
Color Yellow
pH . 7
Boiling Point 332°C
Packaging Glass Jar
Molecular Formula C6H6N2O2
MDL Number MFCD00007858
Quantity 100 g
Synonym p-nitroaniline, p-nitraniline, 4-nitrobenzenamine, p-aminonitrobenzene, 4-nitraniline, benzenamine, 4-nitro, p-nitrophenylamine, 1-amino-4-nitrobenzene, aniline, p-nitro, developer p
InChI Key TYMLOMAKGOJONV-UHFFFAOYSA-N
SMILES NC1=CC=C(C=C1)[N+]([O-])=O
IUPAC Name 4-nitroaniline
Molecular Weight (g/mol) 138.13
PubChem CID 7475
ChEBI CHEBI:17064
Formula Weight 138.13g/mol
Vapor Pressure 1mmHg 142
Physical Form Solid
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'