missing translation for 'onlineSavingsMsg'
Learn More
4-Nitrobenzaldehyde, 99%, Thermo Scientific Chemicals Product Code.: 10255630

4-Nitrobenzaldehyde, 99%, Thermo Scientific Chemicals

Product Code. 10255630
25 g, Plastic bottle
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle
This item is not returnable. View return policy

Product Code. 10255630

Brand: Thermo Scientific Acros 156990250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 555-16-8
Molecular Formula C7H5NO3
Molecular Weight (g/mol) 151.12
MDL Number MFCD00007346
InChI Key BXRFQSNOROATLV-UHFFFAOYSA-N
Synonym p-nitrobenzaldehyde, benzaldehyde, 4-nitro, p-formylnitrobenzene, benzaldehyde, p-nitro, 4-nitrobenzaldehydde, 4-nitro-benzaldehyde, 4-formylnitrobenzene, p-nitro benzaldehyde, 4-nitro benzaldehyde, ccris 1675
PubChem CID 541
ChEBI CHEBI:66926
IUPAC Name 4-nitrobenzaldehyde
SMILES [O-][N+](=O)C1=CC=C(C=O)C=C1

Specifications

Melting Point 105.0°C to 106.0°C
Color Yellow to Brown
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Linear Formula O2NC6H4CHO
Quantity 25 g
Beilstein 07,256
Merck Index 14,6587
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in glacial acetic acid,soluble in alcohol and benzene,slightly soluble in ether
Formula Weight 151.12
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 4-Nitrobenzaldehyde
Show More Show Less
compliance-icons
Product Identifier
  • 4-Nitrobenzaldehyde
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H412-Harmful to aquatic life with long lasting effects.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Nitrobenzaldehyde, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.