missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrobenzeneboronic acid, 95%, Thermo Scientific Chemicals

Product Code. 11322939
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It is reagent used forligand-free palladium-catalyzed Suzuki-Miyaura cross-couplings, ruthenium catalyzed direct arylation of benzylic sp3 carbons of acyclic amines, Diels-Alder or C-H activation reactions, regioselective Suzuki-Miyaura coupling and tandem palladium-catalyzed intramolecular aminocarbonylation and annulations, N-arylation of phenylurea using copper acetylacetonate catalyst, environmentally benign one-pot synthesis through a double arylation process, copper-mediated cyanation, copper-catalyzed arylations regioselective glycosylations. Suzuki couplings followed by arylations, X-ray absorption on rhodium-grafted hydrotalcite catalyst for heterogeneous 1,4-addition reaction of organoboron reagents to electron deficient olefins. It is used in Preparation of combretastatin analogs as potential antitumor agents and human immunodeficiency virus (HIV) protease inhibitors with antiviral activities against drug-resistant viruses

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is reagent used forligand-free palladium-catalyzed Suzuki-Miyaura cross-couplings, ruthenium catalyzed direct arylation of benzylic sp3 carbons of acyclic amines, Diels-Alder or C-H activation reactions, regioselective Suzuki-Miyaura coupling and tandem palladium-catalyzed intramolecular aminocarbonylation and annulations, N-arylation of phenylurea using copper acetylacetonate catalyst, environmentally benign one-pot synthesis through a double arylation process, copper-mediated cyanation, copper-catalyzed arylations regioselective glycosylations. Suzuki couplings followed by arylations, X-ray absorption on rhodium-grafted hydrotalcite catalyst for heterogeneous 1,4-addition reaction of organoboron reagents to electron deficient olefins. It is used in Preparation of combretastatin analogs as potential antitumor agents and human immunodeficiency virus (HIV) protease inhibitors with antiviral activities against drug-resistant viruses

Solubility
Slightly soluble in water.

Notes
Store in cool dry conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24067-17-2
Molecular Formula C6H6BNO4
Molecular Weight (g/mol) 166.93
MDL Number MFCD00161360
InChI Key NSFJAFZHYOAMHL-UHFFFAOYSA-N
Synonym 4-nitrophenyl boronic acid, 4-nitrobenzeneboronic acid, p-nitrophenylboronic acid, 4-nitrophenylboronicacid, p-nitrophenyl boronic acid, 4-nitro phenyl boronic acid, boronic acid, 4-nitrophenyl, boronic acid, b-4-nitrophenyl, 4-borononitrobenzene
PubChem CID 2773552
IUPAC Name (4-nitrophenyl)boronic acid
SMILES OB(O)C1=CC=C(C=C1)[N+]([O-])=O

Specifications

Melting Point ∽285°C (decomposition)
Quantity 5 g
Solubility Information Slightly soluble in water.
Formula Weight 166.93
Percent Purity 95%
Chemical Name or Material 4-Nitrobenzeneboronic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'