Learn More
4-Nitrobenzoic acid, 99+%, Thermo Scientific Chemicals
£14.80 - £50.00
Chemical Identifiers
CAS | 62-23-7 |
---|---|
Molecular Formula | C7H5NO4 |
Molecular Weight (g/mol) | 167.12 |
MDL Number | MFCD00007352 |
InChI Key | OTLNPYWUJOZPPA-UHFFFAOYSA-N |
Synonym | 4-nitrobenzoic acid, p-nitrobenzoic acid, benzoic acid, 4-nitro, nitrodracylic acid, 4-nitrodracylic acid, 1-carboxy-4-nitrobenzene, benzoic acid, p-nitro, p-nitrobenzoicacid, p-nitrobenzenecarboxylic acid, kyselina p-nitrobenzoova |
PubChem CID | 6108 |
ChEBI | CHEBI:262350 |
SMILES | C1=CC(=CC=C1C(=O)O)[N+](=O)[O-] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10376480
|
Thermo Scientific Acros
128460250 |
25 g | Plastic bottle |
£14.80
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10532911
|
Thermo Scientific Acros
128461000 |
100 g | Plastic bottle |
£15.70
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10755051
|
Thermo Scientific Acros
128460010 |
1 kg | Plastic bottle |
£50.00
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
62-23-7 | |
167.12 | |
OTLNPYWUJOZPPA-UHFFFAOYSA-N | |
6108 | |
C1=CC(=CC=C1C(=O)O)[N+](=O)[O-] |
C7H5NO4 | |
MFCD00007352 | |
4-nitrobenzoic acid, p-nitrobenzoic acid, benzoic acid, 4-nitro, nitrodracylic acid, 4-nitrodracylic acid, 1-carboxy-4-nitrobenzene, benzoic acid, p-nitro, p-nitrobenzoicacid, p-nitrobenzenecarboxylic acid, kyselina p-nitrobenzoova | |
CHEBI:262350 |
Specifications
62-23-7 | |
100.0 | |
Yellow | |
99% min. (GC) | |
O2NC6H4CO2H | |
09,389 | |
4-nitrobenzoic acid, p-nitrobenzoic acid, benzoic acid, 4-nitro, nitrodracylic acid, 4-nitrodracylic acid, 1-carboxy-4-nitrobenzene, benzoic acid, p-nitro, p-nitrobenzoicacid, p-nitrobenzenecarboxylic acid, kyselina p-nitrobenzoova | |
OTLNPYWUJOZPPA-UHFFFAOYSA-N | |
167.12 | |
CHEBI:262350 | |
99+% | |
4-Nitrobenzoic acid, 99+% |
99.0 | |
239.0°C to 242.0°C | |
Authentic | |
C7H5NO4 | |
MFCD00007352 | |
14,6589 | |
Solubility in water: insoluble. Other solubilities: 1g/110mL alcohol- 1g/45mL ether,1g/12mL methanol- 1g/20mL acetone,1g/150mL chloroform,slightly soluble in benzene and carbon disulfide | |
C1=CC(=CC=C1C(=O)O)[N+](=O)[O-] | |
6108 | |
167.12 | |
Crystalline Powder |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes serious eye irritation.
GHS P Statement
Wear eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash face,hands and any exposed skin thoroughly after handling.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 200-526-2
missing translation for 'rtecsNumber' : DH5075000
missing translation for 'tsca' : TSCA
- 4-Nitrobenzoic acid
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Carcinogenicity Category 2
- Germ cell mutagenicity Category 2
- Reproductive toxicity Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H319-Causes serious eye irritation.
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H341-Suspected of causing genetic defects.
- H351-Suspected of causing cancer.
- H361-Suspected of damaging fertility or the unborn child.
Precautionary Statement
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only