missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrobenzoic acid, 99%, Thermo Scientific Chemicals

Product Code. 11427786
Click to view available options
Quantity:
250 g
1000 g
5000 g
This item is not returnable. View return policy
This item is not returnable. View return policy

4-Nitrobenzoic acid used as an intermediate for bulk drugs especially in the manufacturing of folic acid and in the manufacturing of PABA, DABA & dye intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Nitrobenzoic acid used as an intermediate for bulk drugs especially in the manufacturing of folic acid and in the manufacturing of PABA, DABA & dye intermediate.

Solubility
Very slightly soluble in cold water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62-23-7
Molecular Formula C7H5NO4
Molecular Weight (g/mol) 167.12
MDL Number MFCD00007352
InChI Key OTLNPYWUJOZPPA-UHFFFAOYSA-N
Synonym 4-nitrobenzoic acid, p-nitrobenzoic acid, benzoic acid, 4-nitro, nitrodracylic acid, 4-nitrodracylic acid, 1-carboxy-4-nitrobenzene, benzoic acid, p-nitro, p-nitrobenzoicacid, p-nitrobenzenecarboxylic acid, kyselina p-nitrobenzoova
PubChem CID 6108
ChEBI CHEBI:262350
SMILES C1=CC(=CC=C1C(=O)O)[N+](=O)[O-]

Specifications

Melting Point 239°C to 241°C
Density 1.61
Quantity 250 g
Beilstein 973593
Merck Index 14,6589
Solubility Information Very slightly soluble in cold water.
Formula Weight 167.12
Percent Purity 99%
Chemical Name or Material 4-Nitrobenzoic acid
compliance-icons compliance-icons
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Germ cell mutagenicity Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H341-Suspected of causing genetic defects.
  • H351-Suspected of causing cancer.
  • H361-Suspected of damaging fertility or the unborn child.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'