missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Nitrobenzoyl chloride, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
250 g
1000 g
Description
4-Nitrobenzoyl chloride is used in the preparation of polysubstituted furanonaphthoquinoines. It is also involved in Michael addition, Henry reaction, O-alkylation and cycloaddition reactions. Further, it is employed as an intermediate in active pharmaceutical ingredient and dyes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Nitrobenzoyl chloride is used in the preparation of polysubstituted furanonaphthoquinoines. It is also involved in Michael addition, Henry reaction, O-alkylation and cycloaddition reactions. Further, it is employed as an intermediate in active pharmaceutical ingredient and dyes.
Solubility
Soluble in terahydrofuran, dichloromethane, chloroform and pyridine.
Notes
Moisture sensitive. Incompatible with water, alcohols, oxidizing agents and strong bases.
4-Nitrobenzoyl chloride is used in the preparation of polysubstituted furanonaphthoquinoines. It is also involved in Michael addition, Henry reaction, O-alkylation and cycloaddition reactions. Further, it is employed as an intermediate in active pharmaceutical ingredient and dyes.
Solubility
Soluble in terahydrofuran, dichloromethane, chloroform and pyridine.
Notes
Moisture sensitive. Incompatible with water, alcohols, oxidizing agents and strong bases.
Specifications
Specifications
Melting Point | 71°C to 74°C |
Density | 1.53 |
Boiling Point | 155°C (15 mmHg) |
Flash Point | 102°C (215°F) |
Odor | Characteristic |
Quantity | 10 g |
UN Number | UN3261 |
Beilstein | 473192 |
Sensitivity | Moisture sensitive |
Merck Index | 14,6590 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'