Learn More
4-Nitrobenzyl bromide, 99%, Thermo Scientific Chemicals
£30.60 - £316.00
Chemical Identifiers
CAS | 100-11-8 |
---|---|
Molecular Formula | C7H6BrNO2 |
Molecular Weight (g/mol) | 216.03 |
MDL Number | MFCD00007373 |
InChI Key | VOLRSQPSJGXRNJ-UHFFFAOYSA-N |
Synonym | 4-nitrobenzyl bromide, 1-bromomethyl-4-nitrobenzene, p-nitrobenzyl bromide, nitrobenzyl bromide, alpha-bromo-4-nitrotoluene, benzene, 1-bromomethyl-4-nitro, p-bromomethyl nitrobenzene, 4-bromomethyl nitrobenzene, alpha-bromo-p-nitrotoluene, 4-nitrobenzylbromide |
PubChem CID | 66011 |
ChEBI | CHEBI:62822 |
IUPAC Name | 1-(bromomethyl)-4-nitrobenzene |
SMILES | [O-][N+](=O)C1=CC=C(CBr)C=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10081390
|
Thermo Scientific Acros
128530250 |
25 g |
£30.60
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10366530
|
Thermo Scientific Acros
128531000 |
100 g |
£81.20
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11934581
|
Thermo Scientific Acros
128535000 |
500 g |
£316.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
100-11-8 | |
216.03 | |
VOLRSQPSJGXRNJ-UHFFFAOYSA-N | |
66011 | |
1-(bromomethyl)-4-nitrobenzene |
C7H6BrNO2 | |
MFCD00007373 | |
4-nitrobenzyl bromide, 1-bromomethyl-4-nitrobenzene, p-nitrobenzyl bromide, nitrobenzyl bromide, alpha-bromo-4-nitrotoluene, benzene, 1-bromomethyl-4-nitro, p-bromomethyl nitrobenzene, 4-bromomethyl nitrobenzene, alpha-bromo-p-nitrotoluene, 4-nitrobenzylbromide | |
CHEBI:62822 | |
[O-][N+](=O)C1=CC=C(CBr)C=C1 |
Specifications
100-11-8 | |
Yellow to Beige | |
Authentic | |
Glass bottle | |
O2NC6H4CH2Br | |
05,334 | |
4-nitrobenzyl bromide, 1-bromomethyl-4-nitrobenzene, p-nitrobenzyl bromide, nitrobenzyl bromide, alpha-bromo-4-nitrotoluene, benzene, 1-bromomethyl-4-nitro, p-bromomethyl nitrobenzene, 4-bromomethyl nitrobenzene, alpha-bromo-p-nitrotoluene, 4-nitrobenzylbromide | |
VOLRSQPSJGXRNJ-UHFFFAOYSA-N | |
1-(bromomethyl)-4-nitrobenzene | |
66011 | |
216.03 | |
Crystalline Powder |
98.0°C to 100.0°C | |
>100°C | |
98.5% min. (GC) | |
C7H6BrNO2 | |
MFCD00007373 | |
01,736; 08,36 | |
Solubility in water: insoluble. Other solubilities: soluble in dmf,isopropyl acetate and acetone | |
[O-][N+](=O)C1=CC=C(CBr)C=C1 | |
216.03 | |
CHEBI:62822 | |
99% | |
4-Nitrobenzyl bromide |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 202-820-6
- 4-Nitrobenzyl bromide
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only