missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrocinnamyl alcohol, 98%

Product Code. 11434123
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11434123

Brand: Thermo Scientific Alfa Aesar A10338.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 1504-63-8 | C9H9NO3 | 179.175 g/mol

4-Nitrocinnamyl alcohol is used in the preparation of 4-nitrocinnamyl chloride by reacting with tosyl chloride (TsCl).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Nitrocinnamyl alcohol is used in the preparation of 4-nitrocinnamyl chloride by reacting with tosyl chloride (TsCl).

Solubility
Soluble in chloroform, dichloromethane and ethyl Acetate.

Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 1504-63-8
Molecular Formula C9H9NO3
Molecular Weight (g/mol) 179.175
MDL Number MFCD00017045
InChI Key LGXXEDSIJZHDBN-OWOJBTEDSA-N
Synonym 4-nitrocinnamyl alcohol, 2e-3-4-nitrophenyl prop-2-en-1-ol, e-3-4-nitrophenyl prop-2-en-1-ol, p-nitrocinnamyl alcohol, 3-4-nitrophenyl prop-2-en-1-ol, 2-propen-1-ol, 3-4-nitrophenyl, 2-propen-1-ol, 3-p-nitrophenyl, 3-p-nitrophenyl allyl alcohol, 3-4-nitrophenyl-2-propenol
PubChem CID 5462894
IUPAC Name (E)-3-(4-nitrophenyl)prop-2-en-1-ol
SMILES C1=CC(=CC=C1C=CCO)[N+](=O)[O-]

Specifica

Melting Point 126°C to 130°C
Quantity 1 g
Beilstein 1948238
Solubility Information Soluble in chloroform,dichloromethane and ethyl Acetate.
Formula Weight 179.18
Percent Purity 98%
Chemical Name or Material 4-Nitrocinnamyl alcohol

RUO – Research Use Only

Correzione del contenuto del prodotto

Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.

Titolo del prodotto

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.

Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato