missing translation for 'onlineSavingsMsg'
Learn More
4-Nitrocinnamyl alcohol, 98%, Thermo Scientific Chemicals Product Code.: 11454123

4-Nitrocinnamyl alcohol, 98%, Thermo Scientific Chemicals

Product Code. 11454123
25 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 11454123

Brand: Thermo Scientific Alfa Aesar A10338.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

4-Nitrocinnamyl alcohol is used in the preparation of 4-nitrocinnamyl chloride by reacting with tosyl chloride (TsCl).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Nitrocinnamyl alcohol is used in the preparation of 4-nitrocinnamyl chloride by reacting with tosyl chloride (TsCl).

Solubility
Soluble in chloroform, dichloromethane and ethyl Acetate.

Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1504-63-8
Molecular Formula C9H9NO3
Molecular Weight (g/mol) 179.175
MDL Number MFCD00017045
InChI Key LGXXEDSIJZHDBN-OWOJBTEDSA-N
Synonym 4-nitrocinnamyl alcohol, 2e-3-4-nitrophenyl prop-2-en-1-ol, e-3-4-nitrophenyl prop-2-en-1-ol, p-nitrocinnamyl alcohol, 3-4-nitrophenyl prop-2-en-1-ol, 2-propen-1-ol, 3-4-nitrophenyl, 2-propen-1-ol, 3-p-nitrophenyl, 3-p-nitrophenyl allyl alcohol, 3-4-nitrophenyl-2-propenolShow MoreShow Less
PubChem CID 5462894
IUPAC Name (E)-3-(4-nitrophenyl)prop-2-en-1-ol
SMILES C1=CC(=CC=C1C=CCO)[N+](=O)[O-]

Specifications

Melting Point 126°C to 130°C
Quantity 25 g
Beilstein 1948238
Solubility Information Soluble in chloroform,dichloromethane and ethyl Acetate.
Formula Weight 179.18
Percent Purity 98%
Chemical Name or Material 4-Nitrocinnamyl alcohol

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10171507 Certificate TypeCertificate of Analysis Date31/08/2022 Catalog NumberA10338.06,A10338.03,A10338.14
Lot Number10171884 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA10338.06,A10338.03,A10338.14
Lot NumberE7932C Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA10338.06,A10338.03,A10338.14
Lot Number10151897 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA10338.06,A10338.03,A10338.14
Lot Number10143476 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA10338.06,A10338.03,A10338.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Nitrocinnamyl alcohol, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.