missing translation for 'onlineSavingsMsg'
Learn More
4-Nitrophenol, 99%, Thermo Scientific Chemicals Product Code.: 10337570

4-Nitrophenol, 99%, Thermo Scientific Chemicals

Product Code. 10337570
100 g, Plastic bottle
Click to view available options
Quantity:
5 g
100 g
250 g
1 kg
5 kg
Packaging:
Glass Bottle
Plastic bottle
Plastic drum
This item is not returnable. View return policy

Product Code. 10337570

Brand: Thermo Scientific Acros 157051000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-02-7
Molecular Formula C6H5NO3
Molecular Weight (g/mol) 139.11
MDL Number MFCD00007331
InChI Key BTJIUGUIPKRLHP-UHFFFAOYSA-N
Synonym p-nitrophenol, phenol, 4-nitro, paranitrophenol, niphen, 4-hydroxynitrobenzene, p-hydroxynitrobenzene, phenol, p-nitro, mononitrophenol, paranitrofenol, paranitrofenolo
PubChem CID 980
ChEBI CHEBI:16836
IUPAC Name 4-nitrophenol
SMILES OC1=CC=C(C=C1)[N+]([O-])=O

Specifications

Melting Point 112°C to 115°C
Color Yellow to Brown
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Linear Formula O2NC6H4OH
Quantity 100 g
Beilstein 06, 226
Fieser 01,743; 02,297
Merck Index 15, 6708
Formula Weight 139.11
Percent Purity ≥98.5% (GC)
Physical Form Crystalline Powder, Crystals or Chunks
Chemical Name or Material 4-Nitrophenol
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0458108 Certificate TypeCertificate of Analysis Date07/12/2023 Catalog Number157052500,157050050,157050051,157050010,157051000
Lot NumberB0149948 Certificate TypeCertificate of Analysis Date14/11/2023 Catalog Number157052500,157050050,157050051,157050010,157051000
Lot NumberA0453503 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number157052500,157050050,157050051,157050010,157051000
Lot NumberA0402504 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number157052500,157050050,157050051,157050010,157051000
Lot NumberA0393846 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number157052500,157050050,157050051,157050010,157051000
compliance-icons compliance-icons
Product Identifier
  • 4-Nitrophenol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H373-May cause damage to organs.
  • H332-Harmful if inhaled.
  • H312-Harmful in contact with skin.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Nitrophenol, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.