missing translation for 'onlineSavingsMsg'
Learn More
4-Nitrophenol, 99%, Thermo Scientific Chemicals Product Code.: 11466967

4-Nitrophenol, 99%, Thermo Scientific Chemicals

Product Code. 11466967
250 g
Click to view available options
Quantity:
250 g
1000 g
5000 g
This item is not returnable. View return policy

Product Code. 11466967

Brand: Thermo Scientific Alfa Aesar A14376.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

4-Nitrophenol is used as a precursor to prepare phenetidine, acetophenetidne and pH indicator. Its carboxylate ester derivatives are involved as an active component for the construction of amide moieties in peptide synthesis. It is used as an intermediate in the synthesis of paracetamol and N-acetyl-p-aminophenol dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Nitrophenol is used as a precursor to prepare phenetidine, acetophenetidne and pH indicator. Its carboxylate ester derivatives are involved as an active component for the construction of amide moieties in peptide synthesis. It is used as an intermediate in the synthesis of paracetamol and N-acetyl-p-aminophenol dyestuffs.

Solubility
Freely soluble in alcohol, chloroform, ether. Freely soluble in acetone. Slightly soluble in water.

Notes
Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-02-7
Molecular Formula C6H5NO3
Molecular Weight (g/mol) 139.11
MDL Number MFCD00007331
InChI Key BTJIUGUIPKRLHP-UHFFFAOYSA-N
Synonym p-nitrophenol, phenol, 4-nitro, paranitrophenol, niphen, 4-hydroxynitrobenzene, p-hydroxynitrobenzene, phenol, p-nitro, mononitrophenol, paranitrofenol, paranitrofenoloShow MoreShow Less
PubChem CID 980
ChEBI CHEBI:16836
IUPAC Name 4-nitrophenol
SMILES OC1=CC=C(C=C1)[N+]([O-])=O

Specifications

Melting Point 110°C to 115°C
Density 1.48
Boiling Point 279°C (decomposition)
Flash Point 169°C (336°F)
Quantity 250 g
UN Number UN1663
Beilstein 1281877
Sensitivity Light sensitive
Merck Index 14,6621
Solubility Information Freely soluble in alcohol,chloroform,ether; Freely soluble in acetone; Slightly soluble in water.
Formula Weight 139.11
Percent Purity 99%
Chemical Name or Material 4-Nitrophenol
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberM29L024 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog NumberA14376.0B,A14376.30,A14376.0I
Lot NumberY20K023 Certificate TypeCertificate of Analysis Date26/11/2024 Catalog NumberA14376.0B,A14376.30,A14376.0I
Lot NumberX21K002 Certificate TypeCertificate of Analysis Date05/11/2024 Catalog NumberA14376.0B,A14376.30,A14376.0I
Lot NumberW24K028 Certificate TypeCertificate of Analysis Date03/10/2024 Catalog NumberA14376.0B,A14376.30,A14376.0I
Lot NumberT31K025 Certificate TypeCertificate of Analysis Date20/08/2024 Catalog NumberA14376.0B,A14376.30,A14376.0I
compliance-icons compliance-icons
Product Identifier
  • 4-Nitrophenol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H301-Toxic if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H373-May cause damage to organs.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
Supplemental information
  • MIXTURE LIST-Contains : p-Nitrophenol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Nitrophenol, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.