missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrophenyl formate, 97%, Thermo Scientific™

Product Code. 11342640
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1865-01-6
Molecular Weight (g/mol) 167.12
InChI Key IEXRKQFZXJSHOB-UHFFFAOYSA-N
PubChem CID 74628
IUPAC Name (4-nitrophenyl) formate
SMILES C1=CC(=CC=C1[N+](=O)[O-])OC=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 66.0°C to 67.0°C
Color White
Elemental Analysis 48.30 to 52.32% C, 8.05 to 8.72% N
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C7H5NO4
Quantity 5 g
Synonym p-nitrophenyl formate, formic acid, 4-nitrophenyl ester, 4-nitrophenyl formate, formic acid, p-nitrophenyl ester, formic acid 4-nitrophenyl ester, acmc-1bty1, formic acid,4-nitrophenyl ester, formic acid 4-nitro-phenyl ester, 4-nitrophenyl formate hplc
Formula Weight 167.12
Percent Purity 97%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 4-Nitrophenyl formate
Show More Show Less
Product Identifier
  • 4-Nitrophenyl formate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'