missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Nitrophenyl isothiocyanate, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
250 g
Description
4-Nitrophenyl isothiocyanate is used as specializing in synthesizing of organic fine chemicals and pharmaceutical intermediates for use in development and production
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Nitrophenyl isothiocyanate is used as specializing in synthesizing of organic fine chemicals and pharmaceutical intermediates for use in development and production
Solubility
It hydrolyzes with water. Soluble in toluene.
Notes
Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
4-Nitrophenyl isothiocyanate is used as specializing in synthesizing of organic fine chemicals and pharmaceutical intermediates for use in development and production
Solubility
It hydrolyzes with water. Soluble in toluene.
Notes
Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
Specifications
Melting Point | 105°C to 110°C |
Boiling Point | 137°C to 138°C (11 mmHg) |
Odor | Irritating |
Quantity | 10 g |
UN Number | UN2923 |
Beilstein | 640027 |
Sensitivity | Moisture sensitive |
Solubility Information | It hydrolyzes with water. Soluble in toluene. |
Formula Weight | 180.19 |
Percent Purity | 97% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'