missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrophenyl phosphate disodium salt hexahydrate, 5mg tablets, Thermo Scientific Chemicals

Product Code. 15460187
Click to view available options
Quantity:
100 Pc.
1000 Pc.
This item is not returnable. View return policy
This item is not returnable. View return policy

Substrate for alkaline and acid phosphatase determinations

4-Nitrophenyl phosphate disodium salt hexahydrate which acts as a chromogenic substrate for the determination of acid and alkaline phosphatases.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Nitrophenyl phosphate disodium salt hexahydrate which acts as a chromogenic substrate for the determination of acid and alkaline phosphatases.

Solubility
It is soluble in water.

Notes
The tablets should be stored -20°C
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4264-83-9
Molecular Formula C6H4NNa2O6P
Molecular Weight (g/mol) 263.05
MDL Number MFCD00007319
InChI Key VIYFPAMJCJLZKD-UHFFFAOYSA-L
Synonym pnpp, disodium 4-nitrophenylphosphate, sodium 4-nitrophenyl phosphate, disodium 4-nitrophenyl phosphate, phosphoric acid, mono 4-nitrophenyl ester, disodium salt, disodium p-nitrophenyl phosphate, phosphoric acid, mono 4-nitrophenyl ester, sodium salt 1:2, p-nitrophenyl phosphate, pnpp liquid substrate system
PubChem CID 77949
IUPAC Name disodium;(4-nitrophenyl) phosphate
SMILES [Na+].[Na+].[O-][N+](=O)C1=CC=C(OP([O-])([O-])=O)C=C1

Specifications

Color Yellow
Quantity 1000 Pc.
Beilstein 5324661
Formula Weight 371.15 (263.05 Anhydrous)
Physical Form Tablets
Chemical Name or Material 4-Nitrophenyl phosphate disodium salt hexahydrate, 5mg tablets

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'