missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrophenyl phosphate disodium salt hexahydrate, 96%

Product Code. 11495053
missing translation for 'orderingAttributeHoverText'
Quantity:
5 g
25 g
100 g
missing translation for 'unitSize'
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11495053

missing translation for 'mfr': Thermo Scientific Alfa Aesar A12310.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 333338-18-4 | C6H4NNa2O6P | 263.05 g/mol

Substrate for alkaline and acid phosphatase determinations4-Nitrophenyl phosphate disodium salt hexahydrate is a chromogenic substrate used in the determination of acid and alkaline phosphatases.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Substrate for alkaline and acid phosphatase determinations4-Nitrophenyl phosphate disodium salt hexahydrate is a chromogenic substrate used in the determination of acid and alkaline phosphatases.

Solubility
Miscible with water. Immiscible with ethanol and acetone.

Notes
Store in a cool place. Incompatible with strong oxidizing agents, strong acids and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 333338-18-4
Molecular Formula C6H4NNa2O6P
Molecular Weight (g/mol) 263.05
MDL Number MFCD00007319
InChI Key VIYFPAMJCJLZKD-UHFFFAOYSA-L
Synonym pnpp, disodium 4-nitrophenylphosphate, sodium 4-nitrophenyl phosphate, disodium 4-nitrophenyl phosphate, phosphoric acid, mono 4-nitrophenyl ester, disodium salt, disodium p-nitrophenyl phosphate, phosphoric acid, mono 4-nitrophenyl ester, sodium salt 1:2, p-nitrophenyl phosphate, pnpp liquid substrate system
PubChem CID 77949
IUPAC Name disodium;(4-nitrophenyl) phosphate
SMILES [Na+].[Na+].[O-][N+](=O)C1=CC=C(OP([O-])([O-])=O)C=C1

Specifications

Melting Point >300°C
Quantity 5 g
Beilstein 5324661
Solubility Information Miscible with water. Immiscible with ethanol and acetone.
Formula Weight 371.15 (263.06 Anhydrous)
Percent Purity 98%
Chemical Name or Material 4-Nitrophenyl phosphate disodium salt hexahydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.