missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrophenyl phosphate, disodium salt, hexahydrate, 98+%

Product Code. 10655562 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 10655562

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

4-Nitrophenyl phosphate, disodium salt, hexahydrate, 98+%, C6H4NNa2O6P.6H2O, CAS Number-333338-18-4 | CAS: 333338-18-4 | C6H4NNa2O6P | 263.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 333338-18-4
Molecular Formula C6H4NNa2O6P
Molecular Weight (g/mol) 263.05
MDL Number MFCD00007319
InChI Key VIYFPAMJCJLZKD-UHFFFAOYSA-L
Synonym pnpp, disodium 4-nitrophenylphosphate, sodium 4-nitrophenyl phosphate, disodium 4-nitrophenyl phosphate, phosphoric acid, mono 4-nitrophenyl ester, disodium salt, disodium p-nitrophenyl phosphate, phosphoric acid, mono 4-nitrophenyl ester, sodium salt 1:2, p-nitrophenyl phosphate, pnpp liquid substrate system
PubChem CID 77949
IUPAC Name disodium;(4-nitrophenyl) phosphate
SMILES [Na+].[Na+].[O-][N+](=O)C1=CC=C(OP([O-])([O-])=O)C=C1

Specifications

Melting Point >300.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 0.1% max. free 4-nitrophenol (HPLC)
Packaging Glass bottle
Linear Formula O2NC6H4OP(O)(ONa)2·6H2O
Quantity 10 g
Beilstein 06,237
Solubility Information (5% in water) Clear
Formula Weight 371.14
Percent Purity 98+%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 4-Nitrophenyl phosphate, disodium salt, hexahydrate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.