missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Nitrophenyl sulfate, potassium salt, 99+%, Thermo Scientific Chemicals
£98.80 - £334.00
Chemical Identifiers
CAS | 6217-68-1 |
---|---|
Molecular Formula | C6H4KNO6S |
Molecular Weight (g/mol) | 257.26 |
MDL Number | MFCD00007466 |
InChI Key | BITVAZYUWRLLCN-UHFFFAOYSA-M |
Synonym | potassium 4-nitrophenyl sulfate, potassium 4-nitrophenyl sulphate, 4-nitrophenyl sulfate potassium salt, potassium p-nitrophenyl sulfate, sulfuric acid, mono 4-nitrophenyl ester, potassium salt, sulfuric acid, mono 4-nitrophenyl ester, potassium salt 1:1, p-nitrophenyl sulfate potassium salt, potassium p-nitrophenyl sulphate, phenol, p-nitro-, hydrogen sulfate ester , potassium salt |
PubChem CID | 80348 |
IUPAC Name | potassium;(4-nitrophenyl) sulfate |
SMILES | [K+].[O-][N+](=O)C1=CC=C(OS([O-])(=O)=O)C=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10542821
|
Thermo Scientific Acros
227490010 |
1 g | Glass bottle |
£98.80
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10791915
|
Thermo Scientific Acros
227490050 |
5 g | Glass bottle |
£334.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
6217-68-1 | |
257.26 | |
BITVAZYUWRLLCN-UHFFFAOYSA-M | |
80348 | |
[K+].[O-][N+](=O)C1=CC=C(OS([O-])(=O)=O)C=C1 |
C6H4KNO6S | |
MFCD00007466 | |
potassium 4-nitrophenyl sulfate, potassium 4-nitrophenyl sulphate, 4-nitrophenyl sulfate potassium salt, potassium p-nitrophenyl sulfate, sulfuric acid, mono 4-nitrophenyl ester, potassium salt, sulfuric acid, mono 4-nitrophenyl ester, potassium salt 1:1, p-nitrophenyl sulfate potassium salt, potassium p-nitrophenyl sulphate, phenol, p-nitro-, hydrogen sulfate ester , potassium salt | |
potassium;(4-nitrophenyl) sulfate |
Specifications
6217-68-1 | |
100.0 | |
Yellow | |
0.05% max. free p-nitrophenol (HPLC) | |
O2NC6H4OSO3K | |
06, II, 225 | |
Solubility in water: slightly soluble in water. | |
[K+].[O-][N+](=O)C1=CC=C(OS([O-])(=O)=O)C=C1 | |
257.26 | |
257.26 | |
Crystalline Powder |
99.0 | |
246.0°C to 250.0°C | |
Authentic | |
C6H4KNO6S | |
MFCD00007466 | |
potassium 4-nitrophenyl sulfate, potassium 4-nitrophenyl sulphate, 4-nitrophenyl sulfate potassium salt, potassium p-nitrophenyl sulfate, sulfuric acid, mono 4-nitrophenyl ester, potassium salt, sulfuric acid, mono 4-nitrophenyl ester, potassium salt 1:1, p-nitrophenyl sulfate potassium salt, potassium p-nitrophenyl sulphate, phenol, p-nitro-, hydrogen sulfate ester , potassium salt | |
BITVAZYUWRLLCN-UHFFFAOYSA-M | |
potassium;(4-nitrophenyl) sulfate | |
80348 | |
99+% | |
4-Nitrophenyl sulfate, potassium salt |
Safety and Handling
missing translation for 'einecsNumber' : 228-288-5
RUO – Research Use Only