missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrophenyl trifluoromethanesulfonate, 99%, Thermo Scientific Chemicals

£19.50 - £58.60

Chemical Identifiers

CAS 17763-80-3
Molecular Formula C7H4F3NO5S
Molecular Weight (g/mol) 271.17
MDL Number MFCD00192354
InChI Key NDTIXHNCNLKURN-UHFFFAOYSA-N
Synonym trifluoromethanesulfonic acid 4-nitrophenyl ester, 4-nitrophenyltriflate, 4-nitrophenyl triflate, acmc-20amc8, 4-nitrophenyl trifluoromethyl sulfonate, 4-nitrophenyl trifluoromethanesulfonate, 1-trifluoromethylsulfonyloxy-4-nitrobenzene, 4-trifluoromethylsulfonyloxy-1-nitrobenzene
PubChem CID 549870
IUPAC Name (4-nitrophenyl) trifluoromethanesulfonate
SMILES [O-][N+](=O)C1=CC=C(OS(=O)(=O)C(F)(F)F)C=C1
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11398446
View Documents
Thermo Scientific Alfa Aesar
L17481.03
1 g
£19.50
1g
Please to purchase this item. Need a web account? Register with us today!
11308456
View Documents
Thermo Scientific Alfa Aesar
L17481.06
5 g
£58.60
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

17763-80-3
271.17
NDTIXHNCNLKURN-UHFFFAOYSA-N
549870
[O-][N+](=O)C1=CC=C(OS(=O)(=O)C(F)(F)F)C=C1
C7H4F3NO5S
MFCD00192354
trifluoromethanesulfonic acid 4-nitrophenyl ester, 4-nitrophenyltriflate, 4-nitrophenyl triflate, acmc-20amc8, 4-nitrophenyl trifluoromethyl sulfonate, 4-nitrophenyl trifluoromethanesulfonate, 1-trifluoromethylsulfonyloxy-4-nitrobenzene, 4-trifluoromethylsulfonyloxy-1-nitrobenzene
(4-nitrophenyl) trifluoromethanesulfonate

Specifications

17763-80-3
C7H4F3NO5S
UN2923
Moisture sensitive
NDTIXHNCNLKURN-UHFFFAOYSA-N
(4-nitrophenyl) trifluoromethanesulfonate
549870
99%
50°C to 54°C
MFCD00192354
1998336
trifluoromethanesulfonic acid 4-nitrophenyl ester, 4-nitrophenyltriflate, 4-nitrophenyl triflate, acmc-20amc8, 4-nitrophenyl trifluoromethyl sulfonate, 4-nitrophenyl trifluoromethanesulfonate, 1-trifluoromethylsulfonyloxy-4-nitrobenzene, 4-trifluoromethylsulfonyloxy-1-nitrobenzene
[O-][N+](=O)C1=CC=C(OS(=O)(=O)C(F)(F)F)C=C1
271.17
271.17
4-Nitrophenyl trifluoromethanesulfonate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.

P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H302+H312+H332-H314-H335

missing translation for 'dotInformation' : Hazard Class: 8

missing translation for 'einecsNumber' : 000-000-0

missing translation for 'tsca' : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.