missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Nitrophenylacetic acid, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
500 g
Description
It is an important raw material and intermediate used in organic synthesis and also used for medicine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is an important raw material and intermediate used in organic synthesis and also used for medicine.
Solubility
Slightly soluble in water.
Notes
Store in cool dry place. Keep away from oxidizing agents.
It is an important raw material and intermediate used in organic synthesis and also used for medicine.
Solubility
Slightly soluble in water.
Notes
Store in cool dry place. Keep away from oxidizing agents.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 152°C to 156°C |
Quantity | 25 g |
Beilstein | 1911801 |
Merck Index | 14,6622 |
Solubility Information | Slightly soluble in water. |
Formula Weight | 181.15 |
Percent Purity | 98% |
Chemical Name or Material | 4-Nitrophenylacetic acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'