missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Nitrostyrene, 98%, Thermo Scientific™
Click to view available options
Quantity:
25 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
Can be used to introduce amino groups into polymers. The thermal degradation of styrene-4-nitrostyrene copolymers (SNS) has been studied using differential thermal analysis (DTA) and thermogravimetry (TGA) under isothermal and dynamic conditions in dynamic nitrogen. 4—Nitrostyrene gives isomeric (4-nitropheny1)-propiona1dehydes in a 96% yield.
Solubility
Insoluble in water.
Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, light, air, heat. Store at 4 deg C.
Can be used to introduce amino groups into polymers. The thermal degradation of styrene-4-nitrostyrene copolymers (SNS) has been studied using differential thermal analysis (DTA) and thermogravimetry (TGA) under isothermal and dynamic conditions in dynamic nitrogen. 4—Nitrostyrene gives isomeric (4-nitropheny1)-propiona1dehydes in a 96% yield.
Solubility
Insoluble in water.
Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, light, air, heat. Store at 4 deg C.
Specifications
Specifications
Melting Point | 25°C to 28°C |
Density | 1.163 |
Boiling Point | 93°C to 96°C (3 mmHg) |
Flash Point | 109°C (228°F) |
Refractive Index | 1.6045 |
Quantity | 25 g |
Sensitivity | Light sensitive |
Solubility Information | Insoluble in water. |
Formula Weight | 149.15 |
Percent Purity | 98% |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'