Learn More
4-Nitrothiophenol, 96%, Thermo Scientific Chemicals
£90.90 - £322.00
Chemical Identifiers
CAS | 1849-36-1 |
---|---|
Molecular Formula | C6H5NO2S |
Molecular Weight (g/mol) | 155.171 |
MDL Number | MFCD00007343 |
InChI Key | AXBVSRMHOPMXBA-UHFFFAOYSA-N |
Synonym | 4-nitrothiophenol, 4-nitrobenzenethiol, p-nitrothiophenol, benzenethiol, 4-nitro, p-nitrophenyl mercaptan, p-nitrobenzenethiol, benzenethiol, p-nitro, 4-nitrobenzene-1-thiol, 4-nitrothiophenolate, 4-nitro-benzenethiol |
PubChem CID | 15809 |
SMILES | C1=CC(=CC=C1[N+](=O)[O-])S |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15455488
|
Thermo Scientific Alfa Aesar
H59240.06 |
5 g |
£90.90
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15465488
|
Thermo Scientific Alfa Aesar
H59240.14 |
25 g |
£322.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.
Solubility
Partly soluble in water and chloroform.
Notes
Recommended storage temperature: 2 - 4°C. Incompatible with oxidizing agents.
Chemical Identifiers
1849-36-1 | |
155.171 | |
AXBVSRMHOPMXBA-UHFFFAOYSA-N | |
15809 |
C6H5NO2S | |
MFCD00007343 | |
4-nitrothiophenol, 4-nitrobenzenethiol, p-nitrothiophenol, benzenethiol, 4-nitro, p-nitrophenyl mercaptan, p-nitrobenzenethiol, benzenethiol, p-nitro, 4-nitrobenzene-1-thiol, 4-nitrothiophenolate, 4-nitro-benzenethiol | |
C1=CC(=CC=C1[N+](=O)[O-])S |
Specifications
1849-36-1 | |
C6H5NO2S | |
606924 | |
Partly soluble in water and chloroform. | |
C1=CC(=CC=C1[N+](=O)[O-])S | |
15809 | |
96% | |
4-Nitrothiophenol |
72°C to 77°C | |
MFCD00007343 | |
4-nitrothiophenol, 4-nitrobenzenethiol, p-nitrothiophenol, benzenethiol, 4-nitro, p-nitrophenyl mercaptan, p-nitrobenzenethiol, benzenethiol, p-nitro, 4-nitrobenzene-1-thiol, 4-nitrothiophenolate, 4-nitro-benzenethiol | |
AXBVSRMHOPMXBA-UHFFFAOYSA-N | |
155.171 | |
155.17 | |
Technical |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 217-436-4
missing translation for 'rtecsNumber' : DC1850000
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only