missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ 4-Nitrotoluene, 99%
Click to view available options
Quantity:
100 g
500 g
Description
Applications
4-Nitrotoluene is used as a basic chemical in the chemical industry for the manufacturing of intermediates.
Solubility
Soluble in water at 20°C 0.26 g/L. Soluble in methanol.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
4-Nitrotoluene is used as a basic chemical in the chemical industry for the manufacturing of intermediates.
Solubility
Soluble in water at 20°C 0.26 g/L. Soluble in methanol.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
Specifications
Melting Point | 50°C to 54°C |
Boiling Point | 238°C to 239°C |
Flash Point | 106°C (222°F) |
Odor | Almond-like |
Quantity | 500 g |
UN Number | UN3446 |
Beilstein | 1906911 |
Merck Index | 14,6650 |
Formula Weight | 137.14 |
Percent Purity | 99% |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'