missing translation for 'onlineSavingsMsg'
Learn More

4-Nitroveratrole, 98+%

Product Code. 11315169
Click to view available options
Quantity:
25 g
50 g
250 g
Unit Size:
250g
25g
50g
This item is not returnable. View return policy

Product Code. 11315169

Brand: Thermo Scientific Alfa Aesar A18264.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 709-09-1 | C8H9NO4 | 183.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 709-09-1
Molecular Formula C8H9NO4
Molecular Weight (g/mol) 183.16
MDL Number MFCD00007238
InChI Key YFWBUVZWCBFSQN-UHFFFAOYSA-N
Synonym 3,4-dimethoxynitrobenzene, 4-nitroveratrole, benzene, 1,2-dimethoxy-4-nitro, 4-nitrcveratrole, 4-nitro-1,2-dimethoxybenzene, benzene,2-dimethoxy-4-nitro, 1,2-dimethoxy-4-nitrobenzol, 3,4-dimethoxylnitrobenzene, 3,4-dimethoxy-1-nitrobenzene
PubChem CID 69728
IUPAC Name 1,2-dimethoxy-4-nitrobenzene
SMILES COC1=CC=C(C=C1OC)[N+]([O-])=O

Specifications

Melting Point 95°C to 98°C
Boiling Point 221°C to 223°C (10 mmHg)
Quantity 25 g
Beilstein 1878614
Formula Weight 183.16
Percent Purity ≥98%
Chemical Name or Material 4-Nitroveratrole
compliance-icons
Product Identifier
  • 4-Nitroveratrole
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : 4-Nitroveratrole

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.