missing translation for 'onlineSavingsMsg'
Learn More
(4-oxo-2,3,4,5-tetrahydro-1,5-benzothiazepin-3-yl)acetic acid, 97%, Thermo Scientific™ Product Code.: 10778625

(4-oxo-2,3,4,5-tetrahydro-1,5-benzothiazepin-3-yl)acetic acid, 97%, Thermo Scientific™

Product Code. 10778625
1 g, Amber Glass Bottle
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy

Product Code. 10778625

Brand: Thermo Scientific Maybridge CC71601DA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 17547-79-4
Molecular Formula C11H11NO3S
Molecular Weight (g/mol) 237.27
MDL Number MFCD01416244
InChI Key VCSCQLDBPVHQPW-UHFFFAOYNA-N
Synonym 4-oxo-2,3,4,5-tetrahydro-1,5-benzothiazepin-3-yl acetic acid, benzo b perhydrothiazepin-4-one-3-acetic acid, 4-oxo-3,5-dihydro-2h-1,5-benzothiazepin-3-yl acetic acid, 1,5-benzothiazepine-3-aceticacid, 2,3,4,5-tetrahydro-4-oxo, 2-4-oxo-2,3,4,5-tetrahydro-1,5-benzothiazepin-3-yl acetic acid, 2-4-oxo-2h,3h,5h-benzo b 1,4-thiazepin-3-yl acetic acid, 2-4-oxo-3,5-dihydro-2h-1,5-benzothiazepin-3-yl acetic acid, 4-oxo-2,3,4,5-tetrahydro-1,5-benzothiazepine-3-acetic acid, 4-oxo-2,3,4,5-tetrahydro-1,5-benzothiazepin-3-yl acetic acid #, 1,5-benzothiazepine-3-acetic acid, 2,3,4,5-tetrahydro-4-oxo
PubChem CID 580528
IUPAC Name 2-(4-oxo-3,5-dihydro-2H-1,5-benzothiazepin-3-yl)acetic acid
SMILES OC(=O)CC1CSC2=CC=CC=C2NC1=O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 237.28
Percent Purity 97%
Chemical Name or Material (4-Oxo-2,3,4,5-tetrahydro-1,5-benzothiazepin-3-yl)acetic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
(4-oxo-2,3,4,5-tetrahydro-1,5-benzothiazepin-3-yl)acetic acid, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.