missing translation for 'onlineSavingsMsg'
Learn More

4-Oxo-4,5,6,7-tetrahydrobenzo[b]furan-3-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10630923
250 mg
Click to view available options
Quantity:
250 mg
1 g
10 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 56671-28-4
Molecular Formula C9H8O4
Molecular Weight (g/mol) 180.159
MDL Number MFCD00052170
InChI Key FABBWECRHZNMDQ-UHFFFAOYSA-N
Synonym 4-oxo-4,5,6,7-tetrahydrobenzofuran-3-carboxylic acid, 4-oxo-4,5,6,7-tetrahydro-1-benzofuran-3-carboxylic acid, 4-oxo-4,5,6,7-tetrahydrobenzo b furan-3-carboxylic acid, maybridge1_007745, 4-oxo-5,6,7-trihydrobenzo 1,2-b furan-3-carboxylic acid, oxotetrahydrobenzofurancarboxylicacid, 4,5,6,7-tetrahydro-4-oxo-3-benzofurancarboxylic acid, 4-oxo-4,5,6,7-tetrahydrocoumarone-3-carboxylic acid, 3-benzofurancarboxylicacid, 4,5,6,7-tetrahydro-4-oxo, 4,5,6,7-tetrahydro-4-oxobenzofuran-3-carboxylic acid
PubChem CID 713937
IUPAC Name 4-oxo-6,7-dihydro-5H-1-benzofuran-3-carboxylic acid
SMILES C1CC2=C(C(=O)C1)C(=CO2)C(=O)O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 180.16
Percent Purity 97%
Chemical Name or Material 4-Oxo-4,5,6,7-tetrahydrobenzo[b]furan-3-carboxylic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Oxo-4,5,6,7-tetrahydrobenzo[b]furan-3-carboxylic acid, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.