missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Oxo-L-proline hydrobromide, 90+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
Description
It is used as an active pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used as an active pharmaceutical intermediate.
Notes
Moisture sensitive. Store away from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
It is used as an active pharmaceutical intermediate.
Notes
Moisture sensitive. Store away from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 134°C to 140°C |
Formula Weight | 210.03 |
Percent Purity | ≥90% |
Quantity | 250 mg |
Optical Rotation | −32° (c=0.5 in Methanol) |
Sensitivity | Moisture sensitive |
Chemical Name or Material | 4-Oxo-L-proline hydrobromide |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'