missing translation for 'onlineSavingsMsg'
Learn More

4-(Pentafluorothio)benzoyl chloride, 97%, Thermo Scientific™

Product Code. 15487527
Click to view available options
Quantity:
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 197384-98-8
Molecular Formula C7H4ClF5OS
Molecular Weight (g/mol) 266.61
MDL Number MFCD03788523
InChI Key NNFDYUZNKYWPGP-UHFFFAOYSA-N
Synonym 4-pentafluorosulfanyl benzoyl chloride, 4-pentafluorothio benzoyl chloride, sulfur, 4-chlorocarbonyl phenyl pentafluoro-, oc-6-21-9ci, 4-pentafluoro-??-sulfanyl benzoyl chloride, 4-pentafluorosulfanylbenzoic chloride, 4-pentafluorosulphanylbenzoyl chloride, 4-pentafluorosulphanyl benzoyl chloride, 4-chlorocarbonyl phenylsulphur pentafluoride, 4-pentafluoro-$l^ 6-sulfanyl benzoyl chloride, 4-pentafluoro-lambda6-sulfanyl benzoyl chloride
PubChem CID 2779215
IUPAC Name 4-(pentafluoro-$l^{6}-sulfanyl)benzoyl chloride
SMILES FS(F)(F)(F)(F)C1=CC=C(C=C1)C(Cl)=O

Specifications

Quantity 5 g
UN Number UN3265
Sensitivity Moisture sensitive
Formula Weight 266.61
Percent Purity 97%
Chemical Name or Material 4-(Pentafluorothio)benzoyl chloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'