missing translation for 'onlineSavingsMsg'
Learn More

4-Pentenoic acid, 98%, Thermo Scientific Chemicals

Product Code. 11421768
Click to view available options
Quantity:
10 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

The sulfonation of 4-pentenoic acid (Allylacetic acid) yields the corresponding γ-lactone. It was used to inhibit fatty acid oxidation in rat heart mitochondria.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The sulfonation of 4-pentenoic acid (Allylacetic acid) yields the corresponding γ-lactone. It was used to inhibit fatty acid oxidation in rat heart mitochondria.

Solubility
Soluble in alcohol, Slightly soluble in water

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 591-80-0
Molecular Formula C5H8O2
Molecular Weight (g/mol) 100.12
MDL Number MFCD00004408
InChI Key HVAMZGADVCBITI-UHFFFAOYSA-N
Synonym 4-pentenoic acid, allylacetic acid, allyl acetic acid, 4 pa, 3-vinylpropionic acid, delta 4-pentenoic acid, unii-d4s77y29fb, fema no. 2843, .delta.4-pentenoic acid, wln: qv3u1
PubChem CID 61138
ChEBI CHEBI:35936
IUPAC Name pent-4-enoic acid
SMILES OC(=O)CCC=C

Specifications

Melting Point -22°C
Density 0.891
Boiling Point 187°C to 189°C
Flash Point 89°C (192°F)
Odor Pungent
Refractive Index 1.4283
Quantity 100 g
UN Number UN3265
Beilstein 1633696
Solubility Information Soluble in alcohol, Slightly soluble in water
Formula Weight 100.12
Percent Purity 98%
Chemical Name or Material 4-Pentenoic acid
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'