missing translation for 'onlineSavingsMsg'
Learn More

4-Pentylbenzoic acid, 97%, Thermo Scientific™

Product Code. 10566353
Click to view available options
Quantity:
1 g
10 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 26311-45-5
Molecular Formula C12H16O2
Molecular Weight (g/mol) 192.258
MDL Number MFCD00002572
InChI Key CWYNKKGQJYAHQG-UHFFFAOYSA-N
Synonym benzoic acid, 4-pentyl, 4-n-pentylbenzoic acid, p-pentylbenzoic acid, 4-amylbenzoic acid, p-amylbenzoic acid, para-pentylbenzoic acid, benzoic acid, p-pentyl, 4-pentyl benzoic acid, p-n-pentylbenzoic acid, 4-pentylbenzoicacid
PubChem CID 33479
IUPAC Name 4-pentylbenzoic acid
SMILES CCCCCC1=CC=C(C=C1)C(=O)O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula CH3(CH2)4C6H4CO2H
Quantity 1 g
Formula Weight 192.26
Percent Purity 97%
Chemical Name or Material 4-Pentylbenzoic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.