missing translation for 'onlineSavingsMsg'
Learn More

4-Pentynoic acid, 95%, Thermo Scientific Chemicals

Catalog Number 10226700
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
6089-09-4
C5H6O2
98.1
MFCD00004407
MLBYLEUJXUBIJJ-UHFFFAOYSA-M
4-pentynoic acid, propargylacetic acid, pent-4-yn-1-oic acid, 4-pentynoicacid, pent4-ynoic acid, zlchem 796, pubchem3491, pent-4-yn-oic acid, acmc-1az7d
22464
pent-4-ynoic acid
C#CCCC(=O)O
This item is not returnable. View return policy
6089-09-4
C5H6O2
98.1
MFCD00004407
MLBYLEUJXUBIJJ-UHFFFAOYSA-M
4-pentynoic acid, propargylacetic acid, pent-4-yn-1-oic acid, 4-pentynoicacid, pent4-ynoic acid, zlchem 796, pubchem3491, pent-4-yn-oic acid, acmc-1az7d
22464
pent-4-ynoic acid
C#CCCC(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 54.0°C to 58.0°C
Color White to Beige
Boiling Point 110.0°C (30.0 mmHg)
Flash Point 75°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula HC≡CCH2CH2CO2H
Quantity 1 g
Beilstein 02, 481
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol,acetone and,methyl-t.-buthylether
Formula Weight 98.1
Percent Purity 95%
Physical Form Crystalline Powder or Flakes
Chemical Name or Material 4-Pentynoic acid, 95%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 4-Pentynoic acid
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'